1,3-Dibutyl-2-thiourea for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: N,N'-DibutylthioureaN,N'-Di-n-butylthiourea1,3-Dibutylthiourea

M-Clarity™ quality level = MQ200

8.20423.0250 8.20423.0250
EM8.20423.0250CS 1324 USD
CA8.20423.0250 EM8.20423.0250
1,3-Dibutyl-2-thiourea for synthesis, Sigma-Aldrich®
1,3-Dibutyl-2-thiourea
Formula: H₃CCH₂CH₂CH₂NHC(S)NHCH₂CH₂CH₂CH₃
MW: 188.34 g/mol
MDL Number: MFCD00004926
CAS Number: 109-46-6

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point 62 - 65 °C
Molar Mass 188.34 g/mol
Bulk Density 500 kg/m3
Vapor Pressure 0.0032 hPa (25 °C)
pH 6 (0.2 g/l, H2O, 20 °C)
Hill Formula C9H20N2S
Chemical Formula (CH3CH2CH2CH2NH)2CS

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR