Tetrabromo-p-benzoquinone for synthesis, Sigma-Aldrich®

Synonyms: Tetrabromo-1,4-benzoquinoneTetrabromoquinone4-Bromanil2,3,5,6-Tetrabromo-1,4-benzoquinone2,3,5,6-Tetrabromocyclohexa-2,5-diene-1,4-dionep-Bromanil

M-Clarity™ quality level = MQ200

8.20180.0005 8.20180.0005
EM8.20180.0005CS 93511.71 USD
CA8.20180.0005 EM8.20180.0005
Tetrabromo-p-benzoquinone for synthesis, Sigma-Aldrich®
Tetrabromo-p-benzoquinone
Formula: C₆Br₄O₂
MW: 423.68 g/mol
MDL Number: MFCD00013785
CAS Number: 488-48-2

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 296 - 298 °C
Molar Mass 423.68 g/mol
Synonyms p-Bromanile
Hill Formula C6Br4O2
Chemical Formula 1-O=C6Br4-4-(=O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR