2,2'-Dihydroxybiphenyl for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
2,2'-Biphenol
, Biphenyl-2,2'-diol
, 2,2-Biphenol
M-Clarity™ quality level = MQ200
8.20154.0050
8.20154.0050
EM8.20154.0050CS
7983
USD
EM8.20154.0050
CA8.20154.0050
2,2'-Dihydroxybiphenyl for synthesis, Sigma-Aldrich®
2,2'-Dihydroxybiphenyl
Formula:
C₁₂H₁₀O₂ MW: 186.21 g/mol |
MDL Number:
MFCD00002210 CAS Number: 1806-29-7 |
Specification Test Results
Solubility | (20 °C) insoluble |
Melting Point | 106 - 109 °C |
Molar Mass | 186.2 g/mol |
Boiling Point | 314 - 316 °C |
Flash Point | 154 °C |
Ignition Point | 324 °C |
Synonyms | o,o'-Biphenol, 2,2'-Dihydroxydiphenyl |
Hill Formula | C12H10O2 |
Chemical Formula | 2-(HO)C6H4C6H4-2-(OH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...