1-Indanone for synthesis, Sigma-Aldrich®

Synonyms: 2,3-Dihydro-1H-inden-1-one

M-Clarity™ quality level = MQ200

8.20699.0025 8.20699.0025
EM8.20699.0025CS 9512.41 USD
EM8.20699.0025 CA8.20699.0025
1-Indanone for synthesis, Sigma-Aldrich®
1-Indanone
Formula: C₉H₈O
MW: 132.16 g/mol
MDL Number: MFCD00003785
CAS Number: 83-33-0

Order Now

Specification Test Results

Solubility 6.5 g/l (20 °C)
Melting Point 37 - 40 °C
Molar Mass 132.16 g/mol
Boiling Point 243 - 245 °C
Flash Point 113 °C
Ignition Point 525 °C
Synonyms α-Hydrindone, 1-Oxoindane
Hill Formula C9H8O

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR