1,3-Indandione for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 1H-Indene-1,3(2H)-dioneIndan-1,3-dione1,3-Indanedione

M-Clarity™ quality level = MQ200

8.20698.0010 8.20698.0010
EM8.20698.0010CS 12364 USD
EM8.20698.0010 CA8.20698.0010
1,3-Indandione for synthesis, Sigma-Aldrich®
1,3-Indandione
Formula: C₉H₆O₂
MW: 146.15 g/mol
MDL Number: MFCD00003779
CAS Number: 606-23-5

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 129 - 132 °C
Molar Mass 146.14 g/mol
Synonyms 1,3-Hydrindenedione, 1,3-Dioxoindane
Hill Formula C9H6O2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR