Potassium perfluorobutane-1-sulfonate for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
Perfluoro-1-butanesulfonic acid potassium salt
, Perfluoro-1-butanesulphonic acid potassium salt
, Potassium Nonafluoro-1-butanesulphonate
, Potassium nonafluoro-1-butanesulphonate
, Potassium nonafluoro-1-butanesulfonate
, Potassium Perfluoro-1-butanesulphonate
, Potassium Nonafluoro-1-butanesulfonate
, Nonafluoro-1-butanesulphonic acid potassium salt
, Potassium Perfluoro-1-butanesulfonate
, Nonafluoro-1-butanesulfonic acid potassium salt
M-Clarity™ quality level = MQ200
8.18662.0010
8.18662.0010
8.18662.0050
EM8.18662.0010CS
60019
USD
CA8.18662.0010
EM8.18662.0010
CA8.18662.0050
Potassium perfluorobutane-1-sulfonate for synthesis, Sigma-Aldrich®
Potassium perfluorobutane-1-sulphonate
Specification Test Results
Solubility | 51 g/l (25 °C) |
Melting Point | 264 - 266 °C |
Molar Mass | 338.19 g/mol |
Bulk Density | 1000 kg/m3 |
pH | 5.5 - 6.5 (50 g/l, H2O) |
Synonyms | Nonafluorobutane-1-sulfonic acid potassium salt |
Hill Formula | C4F9KO3S |
Chemical Formula | CF3(CF2)3SO3K |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...