β-Ionone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: βeta-Jonone4-(2,6,6-Trimethylcyclohex-1-en-1-yl)but-3-en-2-one

M-Clarity™ quality level = MQ200

8.18619.0100 8.18619.0100
EM8.18619.0100CS 1590.96 USD
CA8.18619.0100 EM8.18619.0100
β-Ionone for synthesis, Sigma-Aldrich®
β-Ionone
Formula: C₁₃H₂₀O
MW: 192.30 g/mol
CAS Number: 79-77-6

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point -49 °C
Molar Mass 192.3 g/mol
Boiling Point 127 - 128 °C (13 hPa)
Flash Point 122 °C
Refractive Index 1.5192 (20 °C, 589 nm)
Density 0.94 g/cm3 (20 °C)
Hill Formula C13H20O

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR