4'-Nitrocinnamic acid for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
trans-3-(4-Nitrophenyl)propenoic acid
M-Clarity™ quality level = MQ200
8.18563.0025
8.18563.0025
EM8.18563.0025CS
10785.81
USD
EM8.18563.0025
CA8.18563.0025
4'-Nitrocinnamic acid for synthesis, Sigma-Aldrich®
4'-Nitrocinnamic acid
Formula:
C₉H₇NO₄ MW: 193.15 |
CAS Number:
882-06-4 |
Specification Test Results
Solubility | (20 °C) insoluble |
Melting Point | 288 - 291 °C |
Molar Mass | 193.15 g/mol |
Synonyms | trans-3-(4-Nitrophenyl)propenoic acid |
Hill Formula | C9H7NO4 |
Chemical Formula | 4-(NO2)C6H4CH=CHCOOH |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...