(±)-3-Amino-1,2-propanediol for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
(±)-3-Amino-1,2-dihydroxy propane
, 3-Amino-1,2-propanediol
M-Clarity™ quality level = MQ200
8.18522.0025
8.18522.0025
EM8.18522.0025CS
6853
USD
EM8.18522.0025
CA8.18522.0025
(±)-3-Amino-1,2-propanediol for synthesis, Sigma-Aldrich®
(±)-3-Amino-1,2-propanediol
Formula:
C₃H₉NO₂ MW: 91.11 g/mol |
MDL Number:
MFCD00008140 CAS Number: 616-30-8 |
Specification Test Results
Solubility | (20 °C) soluble |
Molar Mass | 91.11 g/mol |
Boiling Point | 140 - 141 °C (12 hPa) |
Vapor Pressure | 0.01 hPa (20 °C) |
Flash Point | 186 °C |
Refractive Index | 1.492 (20 °C, 589 nm) |
Density | 1.19 g/cm3 (20 °C) |
pH | 11 (100 g/l, H2O, 20 °C) |
Synonyms | 1-Amino-2,3-propanediol |
Hill Formula | C3H9NO2 |
Chemical Formula | NH2CH2CH(OH)CH2OH |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...