2,2'-Methylenebis(6-tert-butyl-p-cresol) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 2,2'-Methylenebis(4-methyl-6-tert-butylphenol)2,2'-Methylenebis(6-tert-butyl-4-methylphenol)2,2-Methylenebis(6-tert-butyl-p-cresol)3,3'-Di-tert-butyl-2,2'-dihydroxy-5,5'-dimethyldiphenylmethaneBis(3-tert-butyl-2-hydroxy-5-methylphenyl)methane

M-Clarity™ quality level = MQ200

8.18447.9045 8.18447.0500
EM8.18447.9045EA 5610 USD
CA8.18447.0500 EM8.18447.9045
2,2'-Methylenebis(6-tert-butyl-p-cresol) for synthesis, Sigma-Aldrich®
2,2'-Methylenebis(6-tert-butyl-p-cresol)
Formula: C₂₃H₃₂O₂
MW: 340.51 g/mol
MDL Number: MFCD00043641
CAS Number: 119-47-1

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point 125 - 128 °C
Molar Mass 340.5 g/mol
Bulk Density 300 kg/m3
Boiling Point 187 °C (0.7 hPa)
Flash Point 195 °C
Density 1.07 g/cm3 (20 °C)
pH 4 - 7 (H2O, 20 °C) (aqueous suspension)
Ignition Point 350 °C
Synonyms 2,2'-Methylenebis-(6-tert-butyl-4-methyl-phenol), Bis(3-tert-butyl-2-hydroxy-5-methylphenyl)-methan
Hill Formula C23H32O2
Chemical Formula (2-(OH)-3-(CH3)3C-5-(CH3)C6H2)2CH2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR