Trifluoromethanesulfonic anhydride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Triflic anhydride

M-Clarity™ quality level = MQ200

8.18043.0050 8.18043.0050
EM8.18043.0050CS 23443 USD
EM8.18043.0050 CA8.18043.0050
Trifluoromethanesulfonic anhydride for synthesis, Sigma-Aldrich®
Trifluoromethanesulphonic anhydride
Formula: C₂F₆O₅S₂
MW: 282.14 g/mol
MDL Number: MFCD00000408
CAS Number: 358-23-6
UN: 3265
ADR: 8,I

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Molar Mass 282.12 g/mol
Boiling Point 82 - 83 °C
Refractive Index 1.3210 - 1.3212 (20 °C, 589 nm)
Density 1.71 g/cm3 (20 °C)
Hill Formula C2F6O5S2
Chemical Formula (CF3SO2)2O

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR