Hexyl butyrate for synthesis, Sigma-Aldrich®

Synonyms: Butyric acid hexyl ester

M-Clarity™ quality level = MQ200

8.14992.0250
CA8.14992.0250
Hexyl butyrate for synthesis, Sigma-Aldrich®
Hexyl butyrate
Formula: C₁₀H₂₀O₂
MW: 172.27 g/mol
MDL Number: MFCD00048884
CAS Number: 2639-63-6

Order Now

Specification Test Results

Melting Point -78 °C
Molar Mass 172.27 g/mol
Boiling Point 208 °C
Flash Point 83 °C
Density 0.865 g/cm3 (20 °C)
Synonyms Butyric acid hexyl ester, Hexyl butanoate
Hill Formula C10H20O2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR