2-(4-Chlorophenylthio)ethanol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 2-[(4-Chlorophenyl)sulphanyl]-1-ethanol4-Chlorophenyl-2-hydroxyethyl sulfide4-Chlorophenyl-2-hydroxyethyl sulphide2-[(4-Chlorophenyl)sulfanyl]-1-ethanol

M-Clarity™ quality level = MQ200

8.14889.0005 8.14889.0005
EM8.14889.0005CS 10245 USD
EM8.14889.0005 CA8.14889.0005
2-(4-Chlorophenylthio)ethanol for synthesis, Sigma-Aldrich®
2-(4-Chlorophenylthio)ethanol
Formula: C₈H₉ClOS
MW: 188.68 g/mol
MDL Number: MFCD00014037
CAS Number: 13457-98-2
ADR: 6.1,III

Order Now

Specification Test Results

Molar Mass 188.68 g/mol
Boiling Point 140 °C (1013 hPa)
Density 1.278 g/cm3 (20 °C)
Hill Formula C8H9ClOS

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR