4-tert-Butylcalix[4]arene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: p-tert-Butylcalix[4]arene

M-Clarity™ quality level = MQ200

8.14707.0001 8.14707.0001
EM8.14707.0001CS 18514 USD
CA8.14707.0001 EM8.14707.0001
4-tert-Butylcalix[4]arene for synthesis, Sigma-Aldrich®
4-tert-Butylcalix[4]arene
Formula: C₄₄H₅₆O₄
MDL Number: MFCD00066280
CAS Number: 60705-62-6

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 344 - 347 °C
Molar Mass 648.92 g/mol
Hill Formula C44H56O4

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR