4,5-Diamino-6-hydroxy-2-pyrimidinethiol hemisulfate-1,5-hydrate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 4(1H)-Pyrimidinone, 5,6-diamino-2,3-dihydro-2-thioxo-, sulfate (2:1)4(1H)-Pyrimidinone, 5,6-diamino-2,3-dihydro-2-thioxo-, sulphate (2:1)Bis(6-amino-4-oxo-2-thioxo-1,2,3,4-tetrahydro-5-pyrimidinaminium) sulphateBis(6-amino-4-oxo-2-thioxo-1,2,3,4-tetrahydro-5-pyrimidinaminium) sulfate

M-Clarity™ quality level = MQ200

8.14702.0005 8.14702.0005
EM8.14702.0005CS 98014 USD
CA8.14702.0005 EM8.14702.0005
4,5-Diamino-6-hydroxy-2-pyrimidinethiol hemisulfate-1,5-hydrate for synthesis, Sigma-Aldrich®
4,5-Diamino-6-hydroxy-2-pyrimidinethiol hemisulfate-1,5-hydrate
Formula: C₈H₁₄N₈O₆S₃
MW: 414.442 g/mol
CAS Number: 207226-25-3

Order Now

Specification Test Results

Melting Point 235 °C
Hill Formula C8H14N8O6S3*(H2O)n

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR