2,5-Dimethylresorcinol for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
2,5-Dimethylbenzene-1,3-diol
M-Clarity™ quality level = MQ200
8.14691.0001
8.14691.0001
EM8.14691.0001CS
125552
USD
CA8.14691.0001
EM8.14691.0001
2,5-Dimethylresorcinol for synthesis, Sigma-Aldrich®
2,5-Dimethylresorcinol
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | 161 - 164 °C |
Molar Mass | 138.17 g/mol |
Boiling Point | 277 - 280 °C |
Synonyms | ß-Orcinol, Dihydroxycylene |
Hill Formula | C8H10O2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...