1-Chloropropane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.18841.0500 8.18841.0100 8.18841.0500 8.18841.0100
EM8.18841.0100CS 1809 USD
EM8.18841.0100 CA8.18841.0500 CA8.18841.0100 EM8.18841.0500
1-Chloropropane for synthesis, Sigma-Aldrich®
1-Chloropropane
Formula: C₃H₇Cl
MW: 78.54 g/mol
MDL Number: MFCD00000995
CAS Number: 540-54-5
UN: 1278
ADR: 3,II

Order Now

Specification Test Results

Solubility 2.7 g/l (20 °C)
Melting Point -122 °C
Molar Mass 78.54 g/mol
Boiling Point 45 - 47 °C
Vapor Pressure 373 hPa (20 °C)
Flash Point -18 °C
Refractive Index 1.388 (20 °C, 589 nm)
Explosion Limit 2.6 - 11.1 %(V)
Density 0.89 g/cm3 (20 °C)
Ignition Point 520 °C
Synonyms Propyl chloride
Hill Formula C3H7Cl

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR