3,3'-Diamino diphenyl sulfone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 3,3'-Sulfonyldianiline3,3'-sulfonyldianiline3-Aminophenyl sulfoneDADPS3-Aminophenyl sulphone3,3'-Sulphonyldianiline3,3'-sulphonyldianiline

M-Clarity™ quality level = MQ200

8.14680.0025 8.14680.0025
EM8.14680.0025CS 1012 USD
CA8.14680.0025 EM8.14680.0025
3,3'-Diamino diphenyl sulfone for synthesis, Sigma-Aldrich®
3,3'-Diamino diphenyl sulphone
Formula: C₁₂H₁₂N₂O₂S
MW: 248.31 g/mol
MDL Number: MFCD00007792
CAS Number: 599-61-1

Order Now

Specification Test Results

Solubility (20 °C) slightly soluble
Melting Point 167 - 170 °C
Molar Mass 248.29 g/mol
Bulk Density 400 kg/m3
Synonyms 3,3'-Sulfonyldianiline
Hill Formula C12H12N2O2S

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR