(-)-[(8,8-Dichlorocamphoryl)sulfonyl]oxaziridine for synthesis, Sigma-Aldrich®

Synonyms: (4aR,7R,8aS)-8,8-Dichloro-9,9-dimethyltetrahydro-4H-4a,7-methanobenzo[c][1,2]oxazireno[2,3-b]isothiazole 3,3-dioxide

M-Clarity™ quality level = MQ200

8.14672.0005 8.14672.0005
EM8.14672.0005CS 142032.74 USD
EM8.14672.0005 CA8.14672.0005
(-)-[(8,8-Dichlorocamphoryl)sulfonyl]oxaziridine for synthesis, Sigma-Aldrich®
(-)-[(8,8-Dichlorocamphoryl)sulfonyl]oxaziridine
Formula: C₁₀H₁₃Cl₂NO₃S
MW: 298.178 g/mol
MDL Number: MFCD00243016
CAS Number: 139628-16-3

Order Now

Specification Test Results

Melting Point 180 - 185 °C
Molar Mass 298.18 g/mol
Hill Formula C10H13Cl2NO3S

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR