Tetrabutylammonium chloride for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.14645.0025 8.14645.0100 8.14645.0100 8.14645.0025
EM8.14645.0100CS 8367 USD
CA8.14645.0100 EM8.14645.0100 EM8.14645.0025 CA8.14645.0025
Tetrabutylammonium chloride for synthesis, Sigma-Aldrich®
Tetrabutylammonium chloride
Formula: C₁₆H₃₆ClN
MW: 277.92 g/mol
MDL Number: MFCD00011635
CAS Number: 1112-67-0

Order Now

Specification Test Results

Melting Point 41 - 44 °C
Molar Mass 277.92 g/mol
Bulk Density 700 kg/m3
Flash Point 110 °C
pH 5 - 8 (100 g/l, H2O, 20 °C)
Hill Formula C16H36ClN

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR