Ethylenediammonium diiodide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Ethylenediamine dihydroiodide

M-Clarity™ quality level = MQ200

8.14619.0025 8.14619.0025
EM8.14619.0025CS 5968 USD
CA8.14619.0025 EM8.14619.0025
Ethylenediammonium diiodide for synthesis, Sigma-Aldrich®
Ethylenediammonium diiodide
Formula: C₂H₁₀I₂N₂
MW: 315.92 g/mol
MDL Number: MFCD00035535
CAS Number: 5700-49-2

Order Now

Specification Test Results

Solubility (20 °C) soluble
Molar Mass 315.92 g/mol
pH 4.0 - 6.0 (100 g/l, H2O, 20 °C)
Hill Formula C2H10I2N2
Chemical Formula C2H8N2*2HI

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR