8-Hydroxyquinoline-5-sulfonic acid monohydrate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 8-Hydroxyquinoline-5-sulphonic acid hydrate8-Hydroxyquinoline-5-sulfonic acid hydrate

M-Clarity™ quality level = MQ200

8.14436.0100 8.14436.0100
EM8.14436.0100CS 1412 USD
EM8.14436.0100 CA8.14436.0100
8-Hydroxyquinoline-5-sulfonic acid monohydrate for synthesis, Sigma-Aldrich®
8-Hydroxyquinoline-5-sulphonic acid monohydrate
Formula: C₉H₇NO₄S·1H₂O
MW: 243.2365 g/mol
MDL Number: MFCD00149482
CAS Number: 283158-18-9

Order Now

Specification Test Results

Solubility (20 °C) slightly soluble
Melting Point 322 °C
Molar Mass 243.24 g/mol
Hill Formula C9H7NO4S*H2O

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR