α-(Phenylthio)phenylacetic acid for synthesis, Sigma-Aldrich®


M-Clarity™ quality level = MQ200

8.14314.0100 8.14314.0100
EM8.14314.0100CS 1114.4 USD
CA8.14314.0100 EM8.14314.0100
α-(Phenylthio)phenylacetic acid for synthesis, Sigma-Aldrich®
α-(Phenylthio)phenylacetic acid
Formula: C₁₄H₁₂O₂S
MDL Number: MFCD00066259
CAS Number: 10490-07-0

Order Now

Specification Test Results

Melting Point 99 - 101 °C
Molar Mass 244.3 g/mol
Synonyms 2-Phenylthiophenylacetic acid
Hill Formula C14H12O2S

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR