tert-Butyl (2-nitroethyl)ether for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
M-Clarity™ quality level = MQ200
8.14278.0001
8.14278.0001
EM8.14278.0001CS
10670
USD
EM8.14278.0001
CA8.14278.0001
tert-Butyl (2-nitroethyl)ether for synthesis, Sigma-Aldrich®
tert-Butyl (2-nitroethyl) ether
Specification Test Results
Solubility | (20 °C) insoluble |
Molar Mass | 147.17 g/mol |
Boiling Point | 30 - 35 °C (1 hPa) |
Flash Point | 75 °C |
Density | 1.00 g/cm3 (20 °C) |
Synonyms | 2-Nitroethanol tert-butyl ether |
Hill Formula | C6H13NO3 |
Chemical Formula | O2NCH2CH2OC(CH3)3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...