5,5-Dimethyl-2-nitromethyl-1,3-dioxane for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
M-Clarity™ quality level = MQ200
8.14204.0001
8.14204.0001
EM8.14204.0001CS
9963
USD
EM8.14204.0001
CA8.14204.0001
5,5-Dimethyl-2-nitromethyl-1,3-dioxane for synthesis, Sigma-Aldrich®
5,5-Dimethyl-2-nitromethyl-1,3-dioxane
Specification Test Results
Solubility | (20 °C) insoluble |
Molar Mass | 175.18 g/mol |
Boiling Point | 67 - 69 °C (1 hPa) |
Density | 1.14 g/cm3 (20 °C) |
Hill Formula | C7H13NO4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...