1,1-Dimethoxy-3-nitropropane for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
M-Clarity™ quality level = MQ200
8.14137.0001
8.14137.0001
EM8.14137.0001CS
10056
USD
EM8.14137.0001
CA8.14137.0001
1,1-Dimethoxy-3-nitropropane for synthesis, Sigma-Aldrich®
1,1-Dimethoxy-3-nitropropane
Formula:
C₅H₁₁NO₄ |
CAS Number:
72447-81-5 |
Specification Test Results
Solubility | (20 °C) insoluble |
Molar Mass | 149.14 g/mol |
Boiling Point | 96 °C (20 hPa) |
Flash Point | 90 °C |
Density | 1.12 g/cm3 (20 °C) |
Synonyms | 3-Nitropropionaldehyde dimethyl acetal |
Hill Formula | C5H11NO4 |
Chemical Formula | O2N-CH2-CH2-CH(OCH3)2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...