1,10-Didecyl-1,10-diaza-18-crown-6 (7,16-didecyl-1,4,10,13-tetraoxa-7,16-diazacyclooctadecane kryptofix® 22 DD) for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
M-Clarity™ quality level = MQ200
8.11963.0001
8.11963.0001
EM8.11963.0001CS
821780
USD
EM8.11963.0001
CA8.11963.0001
1,10-Didecyl-1,10-diaza-18-crown-6 (7,16-didecyl-1,4,10,13-tetraoxa-7,16-diazacyclooctadecane kryptofix® 22 DD) for synthesis, Sigma-Aldrich®
1,10-Didecyl-1,10-diaza-18-crown-6, (7,16-Didecyl-1,4,10,13-tetraoxa-7,16-diazacyclooctadecane, Kryptofix® 22 DD)
Specification Test Results
Solubility | (20 °C) insoluble |
Melting Point | 36 - 39 °C |
Molar Mass | 542.88 g/mol |
Synonyms | 4,13-Didecyl-1,7,10,16-tetraoxa-4,13-diazacyclooctadecane |
Hill Formula | C32H66N2O4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...