2,2,2-Trichloroethanol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 2,2,2-Trichloroethan-1-ol

M-Clarity™ quality level = MQ200

8.08610.0100
EM8.08610.0100CS 2435 USD
EM8.08610.0100
2,2,2-Trichloroethanol for synthesis, Sigma-Aldrich®
2,2,2-Trichloroethanol
Formula: C₂H₃Cl₃O
MW: 149.4 g/mol
MDL Number: MFCD00004677
CAS Number: 115-20-8
ADR: 6.1,III

Order Now

Specification Test Results

Solubility 83 g/l (20 °C)
Melting Point 17 - 18 °C
Molar Mass 149.4 g/mol
Boiling Point 151 - 153 °C (1013 hPa)
Vapor Pressure 1.3 hPa (20 °C)
Flash Point 110 °C
Refractive Index 1.4885 (20 °C, 589 nm)
Density 1.56 g/cm3 (20 °C)
pH 5 - 6 (H2O, 20 °C) (saturated solution)
Ignition Point 151 °C
Synonyms ß,ß,ß-Trichloroethyl alcohol
Hill Formula C2H3Cl3O
Chemical Formula CCl3CH2OH

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR