1-Undecanol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 1-Undecyl alcoholDecyl carbinolUndecan-1-olUndecyl alcohol1-Hydroxy undecane

M-Clarity™ quality level = MQ200

8.08468.0100 8.08468.0100
EM8.08468.0100CS 1088 USD
EM8.08468.0100 CA8.08468.0100
1-Undecanol for synthesis, Sigma-Aldrich®
1-Undecanol
Formula: C₁₁H₂₄O
MW: 172.31 g/mol
MDL Number: MFCD00004751
CAS Number: 112-42-5
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 13 - 15 °C
Molar Mass 172.31 g/mol
Boiling Point 129 - 131 °C (16 hPa)
Flash Point 113 °C
Refractive Index 1.44 (20 °C, 589 nm)
Density 0.83 g/cm3 (20 °C)
pH 7 (H2O, 20 °C)
Synonyms Undecyl alcohol
Hill Formula C11H24O
Chemical Formula CH3(CH2)10OH

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR