(S)-(+)-2-Amino-3-methyl-1-butanol for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: L-ValinolL(+)-ValinolL(+)-2-Amino-3-methyl-1-butanol(S)-2-Amino-3-methylbutan-1-ol(S)-2-Amino-3-methyl-butan-1-ol

M-Clarity™ quality level = MQ200

8.14082.0001 8.14082.0001
EM8.14082.0001CS 2555 USD
EM8.14082.0001 CA8.14082.0001
(S)-(+)-2-Amino-3-methyl-1-butanol for synthesis, Sigma-Aldrich®
(S)-(+)-2-Amino-3-methyl-1-butanol
Formula: C₅H₁₃NO
MW: 103.16 g/mol
MDL Number: MFCD00064296
CAS Number: 2026-48-4
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 31 - 32 °C
Molar Mass 103.16 g/mol
Boiling Point 91 - 92 °C (13 hPa)
Flash Point 78 °C
Refractive Index 1.4548 (20 °C, 589 nm)
Density 0.93 g/cm3 (20 °C)
Synonyms L-Valinol
Hill Formula C5H13NO
Chemical Formula CH3(CH3)CHCH(NH2)CH2OH

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR