Hydantoin for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
Imidazolidine-2,4-dione
M-Clarity™ quality level = MQ200
8.04511.0100
8.04511.0500
8.04511.0500
8.04511.0100
EM8.04511.0100CS
7777
USD
EM8.04511.0100
CA8.04511.0500
EM8.04511.0500
CA8.04511.0100
Hydantoin for synthesis, Sigma-Aldrich®
Hydantoin
Formula:
C₃H₄N₂O₂ MW: 100.08 g/mol |
MDL Number:
MFCD00005259 CAS Number: 461-72-3 |
Specification Test Results
Solubility | (20 °C) slightly soluble |
Melting Point | 219 - 222 °C |
Molar Mass | 100.08 g/mol |
Synonyms | 2,4-(3H,5H)-Imidazoledione |
Hill Formula | C3H4N2O2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...