2'-Hydroxyacetophenone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 1-(2-Hydroxyphenyl)ethan-1-oneo-Acetylphenol1-Ethanoyl-2-hydroxybenzolo-Hydroxyacetophenone2-Acetylphenol1-(2-Hydroxyphenyl)-1-oxoethane2-Hydroxyphenyl Methyl Ketone2-Acetophenol

M-Clarity™ quality level = MQ200

8.04310.0250 8.04310.0250
EM8.04310.0250CS 1480 USD
CA8.04310.0250 EM8.04310.0250
2'-Hydroxyacetophenone for synthesis, Sigma-Aldrich®
2'-Hydroxyacetophenone
Formula: C₈H₈O₂
MW: 136.15 g/mol
MDL Number: MFCD00002219
CAS Number: 118-93-4

Order Now

Specification Test Results

Solubility 0.2 g/l (20 °C)
Melting Point 4 - 6 °C
Molar Mass 136.15 g/mol
Boiling Point 212 - 214 °C (1013 hPa)
Vapor Pressure 0.27 hPa (20 °C)
Flash Point 106 °C
Refractive Index 1.5584 (20 °C, 589 nm)
Explosion Limit 0.98 - 11.8 %(V)
Density 1.13 g/cm3 (20 °C)
Synonyms 2-Acetylphenol
Hill Formula C8H8O2
Chemical Formula 2-(OH)C6H4COCH3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR