Glutardialdehyde bis(sodium hydrogen sulfite) for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
M-Clarity™ quality level = MQ200
8.04240.0250
8.04240.0250
EM8.04240.0250CS
1885
USD
CA8.04240.0250
EM8.04240.0250
Glutardialdehyde bis(sodium hydrogen sulfite) for synthesis, Sigma-Aldrich®
Glutardialdehyde bis(sodium hydrogen sulphite)
Specification Test Results
Solubility | 285 g/l (20 °C) |
Molar Mass | 308.22 g/mol |
pH | 3 - 5 (100 g/l, H2O, 20 °C) |
Synonyms | Pentane-1,5-dial-bis(sodium bisulfit) |
Hill Formula | C5H10Na2O8S2 |
Chemical Formula | CH2[CH2CH(OH)SO3Na]2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...