Dodecylamine for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Dodecan-1-amine
, 1-Aminododecane
, 1-Dodecylamine
M-Clarity™ quality level = MQ200
8.03527.0101
8.03527.0500
8.03527.0101
8.03527.0500
EM8.03527.0500CS
669
USD
EM8.03527.0500
CA8.03527.0101
CA8.03527.0500
EM8.03527.0101
Dodecylamine for synthesis, Sigma-Aldrich®
Dodecylamine
Formula:
H₃C(CH₂)₁₀CH₂NH₂ MW: 185.35 g/mol |
MDL Number:
MFCD00008154 CAS Number: 124-22-1 UN: 3259 ADR: 8,II |
Specification Test Results
Solubility | (20 °C) practically insoluble |
Melting Point | 25 - 28 °C |
Molar Mass | 185.35 g/mol |
Boiling Point | 247 - 249 °C (1013 hPa) |
Vapor Pressure |
|
Flash Point |
|
Refractive Index | 1.4377 (30 °C) |
Density | 0.80 g/cm3 (20 °C) |
Synonyms | Laurylamine, 1-Aminododecan |
Hill Formula | C12H27N |
Chemical Formula | CH3(CH2)11NH2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...