Cyclohexylamine for synthesis, Sigma-Aldrich®

Danger

Synonyms: Aminocyclohexane

M-Clarity™ quality level = MQ200

8.02885.1000 8.02885.1000 8.02885.0100 8.02885.2500 8.02885.2500
EM8.02885.2500CS 531.65 USD
CA8.02885.2500 CA8.02885.0100 CA8.02885.1000 EM8.02885.2500 EM8.02885.1000
Cyclohexylamine for synthesis, Sigma-Aldrich®
Cyclohexylamine
Formula: C₆H₁₃N
MW: 99.18 g/mol
MDL Number: MFCD00001486
CAS Number: 108-91-8
UN: 2357
ADR: 8 (3),II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -18 °C
Molar Mass 99.17 g/mol
Boiling Point 133 - 134 °C (1013 hPa)
Vapor Pressure 14 hPa (20 °C)
Flash Point 27 °C
Refractive Index 1.458 (20 °C, 589 nm)
Explosion Limit 1.6 - 9.4 %(V)
Density 0.87 g/cm3 (20 °C)
pH 11.5 (100 g/l, H2O, 20 °C)
Ignition Point 265 °C
Synonyms Aminocyclohexane, Cyclohexanamine
Hill Formula C6H13N

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR