Cystamine dihydrochloride for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Bis(2-aminoethyl) disulfide dihydrochloride
, 2,2'-Disulfanediyldiethanamine dihydrochloride
, 2,2′-Diaminodiethyl disulphide dihydrochloride
, Bis(2-aminoethyl) disulphide dihydrochloride
, {2-[(2-Aminoethyl)disulfanyl]ethyl}amine dihydrochloride
, 2,2′-Diaminodiethyl disulfide dihydrochloride
, Cystaminium dichloride
, 2,2′-Dithiobis(ethylamine) dihydrochloride
, {2-[(2-Aminoethyl)disulphanyl]ethyl}amine dihydrochloride
, 2,2'-Disulphanediyldiethanamine dihydrochloride
, Decarboxycystine dihydrochloride
M-Clarity™ quality level = MQ200
Formula:
C₄H₁₂N₂S₂·2HCl MW: 225.20 g/mol |
MDL Number:
MFCD00012905 CAS Number: 56-17-7 |
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | 218 - 221 °C |
Molar Mass | 225.19 g/mol |
Synonyms | Cystamine dihydrochloride |
Hill Formula | C4H14Cl2N2S2 |
Chemical Formula | [H2NCH2CH2SSCH2CH2NH2] * 2 HCl |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...