Cyclopentanone for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
M-Clarity™ quality level = MQ200
8.02670.1000
8.02670.9025
8.02670.0250
8.02670.1000
8.02670.2500
8.02670.0250
8.02670.2500
EM8.02670.9025EA
1679
USD
EM8.02670.9025
CA8.02670.1000
EM8.02670.2500
CA8.02670.0250
EM8.02670.0250
EM8.02670.1000
CA8.02670.2500
Cyclopentanone for synthesis, Sigma-Aldrich®
Cyclopentanone
Formula:
C₅H₈O MW: 84.12 g/mol |
MDL Number:
MFCD00001409 CAS Number: 120-92-3 UN: 2245 ADR: 3,III |
Specification Test Results
Solubility | (20 °C) slightly soluble |
Melting Point | -58 °C |
Molar Mass | 84.11 g/mol |
Boiling Point | 131 °C (1013 hPa) |
Vapor Pressure | 11.5 hPa (20 °C) |
Flash Point | 27 °C |
Explosion Limit | 1.6 - 10.8 %(V) |
Density | 0.95 g/cm3 (20 °C) |
pH | (H2O) not applicable |
Ignition Point | 445 °C |
Hill Formula | C5H8O |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...