Biphenyl-2,2'-dicarboxylic acid for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
Diphenic acid
, [1,1'-Biphenyl]-2,2'-dicarboxylic acid
, 2,2'-Biphenyldicarboxylic acid
M-Clarity™ quality level = MQ200
8.02256.0050
8.02256.0050
EM8.02256.0050CS
21716
USD
EM8.02256.0050
CA8.02256.0050
Biphenyl-2,2'-dicarboxylic acid for synthesis, Sigma-Aldrich®
Biphenyl-2,2'-dicarboxylic acid
Formula:
C₁₄H₁₀O₄ MW: 242.23 g/mol |
MDL Number:
MFCD00002464 CAS Number: 482-05-3 |
Specification Test Results
Solubility | (20 °C) insoluble |
Melting Point | 225 - 228 °C |
Molar Mass | 242.23 g/mol |
Bulk Density | 470 kg/m3 |
pH | 4 - 5 (H2O, 20 °C) |
Ignition Point | 390 °C |
Synonyms | Diphenic acid, Diphenyl-2,2-dicarboxylic acid |
Hill Formula | C14H10O4 |
Chemical Formula | 2-(HOOC)C6H4C6H4-2-(COOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...