Diethyl ethylmalonate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.00906.0250 8.00906.0250
EM8.00906.0250CS 1020 USD
CA8.00906.0250 EM8.00906.0250
Diethyl ethylmalonate for synthesis, Sigma-Aldrich®
Diethyl ethylmalonate
Formula: C₉H₁₆O₄
MDL Number: MFCD00009167
CAS Number: 133-13-1

Order Now

Specification Test Results

Solubility 2.87 g/l (20 °C)
Molar Mass 188.22 g/mol
Boiling Point 204 - 206 °C (1013 hPa)
Flash Point 88 °C
Refractive Index 1.4158 (20 °C, 589 nm)
Density 1.00 g/cm3 (20 °C)
Synonyms Ethylmalonic acid diethyl ester
Hill Formula C9H16O4
Chemical Formula C2H5CH(COOC2H5)2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR