Hexacarbonylmolybdenum for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Molybdenum carbonyl
, Molybdenum hexacarbonyl
, Hexacarbonylmolybdenum(0)
M-Clarity™ quality level = MQ200
8.00396.0010
8.00396.0025
8.00396.0010
8.00396.0025
EM8.00396.0010CS
160704
USD
CA8.00396.0010
CA8.00396.0025
EM8.00396.0010
EM8.00396.0025
Hexacarbonylmolybdenum for synthesis, Sigma-Aldrich®
Hexacarbonylmolybdenum
Formula:
C₆MoO₆ MW: 264 g/mol |
MDL Number:
MFCD00003466 CAS Number: 13939-06-5 UN: 3466 ADR: 6.1,III |
Specification Test Results
Solubility | (20 °C) insoluble |
Melting Point | 147 - 152 °C (decomposition) |
Molar Mass | 264 g/mol |
Boiling Point | 156.4 °C |
Vapor Pressure | 0.13 hPa (20 °C) |
Density | 1.96 g/cm3 (20 °C) |
Synonyms | Molybdenum carbonyl |
Hill Formula | C6MoO6 |
Chemical Formula | Mo(CO)6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...