Tantalum ≥98%, powder, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

1.12349.0025 1.12349.0025
EM1.12349.0025CS 36789 USD
EM1.12349.0025 CA1.12349.0025
Tantalum ≥98%, powder, Sigma-Aldrich®
Tantalum
Formula: Ta
MDL Number: MFCD00011252
CAS Number: 7440-25-7
UN: 3089
ADR: 4.1,

Order Now

Specification Test Results

Solubility  - 0.010 g/l (20 °C)
Melting Point 2996 °C
Molar Mass 180.94 g/mol
Bulk Density 500 kg/m3
Boiling Point 5425 °C
Density 16.6 g/cm3 (20 °C)
pH 7 (H2O, 20 °C) (aqueous suspension)
Ignition Point 250 °C
Hill Formula Ta

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR