Tissue-Tek Cryo Console Plus, 115 V
Supplier: Sakura
5250
10204-848EA 12859.39 USD
10204-848
Tissue-Tek Cryo Console Plus, 115 V
Block Heaters Block Heater/Coolers
Stand-alone cold plate to cool and solidify paraffin blocks in tissue embedding
  • 115 VAC, 60 Hz
  • Control panel allows adjustment of temperature and monitor of the plate from -10 to 0 °C
  • Holds up to 60 blocks