Human Recombinant BMP2 (from E. coli)
Supplier: Tonbo Biosciences
21-8507-U002 21-8507-U010 21-8507-U100
10002-146EA 865.23 USD
10002-146 10273-362 10273-360
Human Recombinant BMP2 (from E. coli)
Bone Morphogenetic Proteins (BMPs) belong to the TGF-beta superfamily of proteins. BMPs elicit their effects by signaling through serine/threonine kinase receptors and play key roles in the development of nearly all organs and tissues. BMP-2 plays a role in the development of bone and cartilage and is also involved n cardiac cell differentiation.
  • Carrier-free