Anti-PRR13 Rabbit Polyclonal Antibody

Supplier: Proteintech
14101-1-AP-150UL
10156-224EA 459.7 USD
10156-224
Anti-PRR13 Rabbit Polyclonal Antibody
Antibodies
The PRR13 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human PRR13. This antibody recognizes human antigen. The PRR13 antibody has been validated for the following applications: ELISA, IHC, WB analysis.

WB 1 : 1000 ; IHC 1 : 200

Type: Primary
Antigen: PRR13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR