Anti-CBX4 Rabbit Polyclonal Antibody

Supplier: Proteintech
18544-1-AP-150UL
10154-830EA 459.7 USD
10154-830
Anti-CBX4 Rabbit Polyclonal Antibody
Antibodies
The CBX4 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human CBX4. This antibody recognizes human,mouse,rat antigen. The CBX4 antibody has been validated for the following applications: ELISA, IF, WB analysis.

WB 1 : 600

Type: Primary
Antigen: CBX4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR