Anti-CNNM4 Rabbit Polyclonal Antibody

Supplier: Proteintech
14066-1-AP-150UL
10156-222EA 459.7 USD
10156-222
Anti-CNNM4 Rabbit Polyclonal Antibody
Antibodies
The CNNM4 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human CNNM4. This antibody recognizes human,mouse,rat antigen. The CNNM4 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 1000 ; IF 1 : 50 ; IHC 1 : 50

Type: Primary
Antigen: CNNM4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR