Anti-IGFBP4 Rabbit Polyclonal Antibody

Supplier: Proteintech
18500-1-AP-150UL
10154-826EA 459.7 USD
10154-826
Anti-IGFBP4 Rabbit Polyclonal Antibody
Antibodies
The IGFBP4 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human IGFBP4. This antibody recognizes human, rat antigen. The IGFBP4 antibody has been validated for the following applications: ELISA, WB, IF, IP analysis.

WB 1 : 300 ; IHC 1 : 50 ; IF 1 : 25

Type: Primary
Antigen: IGFBP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR