Anti-EPAC2 Rabbit Polyclonal Antibody

Supplier: Proteintech
13882-1-AP
10154-532EA 435.29 USD
10154-532
Anti-EPAC2 Rabbit Polyclonal Antibody
Antibodies
The EPAC2 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human EPAC2. This antibody recognizes human,mouse,rat antigen. The EPAC2 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 1000

Type: Primary
Antigen: EPAC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR