Anti-IgJ Rabbit Polyclonal Antibody

Supplier: Proteintech
13688-1-AP-150UL
10156-010EA 459.7 USD
10156-010
Anti-IgJ Rabbit Polyclonal Antibody
Antibodies
The IGJ antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human IGJ. This antibody recognizes human,mouse,rat antigen. The IGJ antibody has been validated for the following applications: ELISA, WB, IHC analysis.

WB 1 : 1000 ; IHC 1 : 100 ; IF 1 : 50

Type: Primary
Antigen: IgJ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR