E-Grade® Function Upgrade, Pilot ONE® Circulator Controller, Huber
Supplier: Huber
9495 9496
10221-690EA 1218.76 USD
10221-690 10221-692
E-Grade® Function Upgrade, Pilot ONE® Circulator Controller, Huber
Circulating Thermostats Open Circulator Systems Thermostat Controllers
The Pilot ONE® controllers can be upgraded giving post sale flexibility by means of their easy to use “Plug and Play” technology
Pilot ONE® Exclusive provides additional features such as Graphical display and display resolution of 0.1 and 0.01 °C, Three programs each with five steps and compact or full menu display, Ramp functionality, True Adaptive Control (TAC) and External control capability.
Pilot ONE® Professional provides additional features such as Non-linear ramping, Ten programs, with a total of 100 program steps, User and administrator menus, Second set point function, and Option for external temperature control via an external PT100 sensor.
“Basic” Pilot ONE® controllers which can be upgraded to Pilot ONE® “Exclusive” or Pilot ONE® “Professional” by simply entering machine specific activation code.
Accessories information: The Profibus gateway enables the connection of Huber temperature control machines to Profibus systems, offering comprehensive facilities for data communication within SPS and process control systems.